Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)cyclohex-1-enylboronic acid pinacol ester, 95%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC434280050

 View more versions of this product

Catalog No. AC434280050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94.0% (GC)
Physical Form Crystalline Powder
Formula Weight 276.11
Quantity 5g
Color White
Melting Point 78° to 83°C
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C13H20BF3O2
CAS 683242-93-5
Chemical Name or Material 4-(Trifluoromethyl)cyclohex-1-enylboronic acid pinacol ester
Assay Percent Range 95%
Synonym 4,4,5,5-Tetramethyl-2-[4-(trifluoromethyl)-1-cyclohexen-1-yl]-1,3,2-dioxaborolane

Specifications

Chemical Name or Material 4-(Trifluoromethyl)cyclohex-1-enylboronic acid pinacol ester
CAS 683242-93-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0% (GC)
Assay Percent Range 95%
Molecular Formula C13H20BF3O2
Formula Weight 276.11
Synonym 4,4,5,5-Tetramethyl-2-[4-(trifluoromethyl)-1-cyclohexen-1-yl]-1,3,2-dioxaborolane
Accessories

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents