Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Trifluoromethyl)biphenyl-2-carboxylic acid, 98%

$203.42 - $800.09

Chemical Identifiers

Chemical Name or Material 4'-(Trifluoromethyl)biphenyl-2-carboxylic acid
CAS 84392-17-6
Molecular Formula C14H9F3O2
Formula Weight 266.22
MDL Number MFCD00075353
Beilstein 7577560
Synonym 4-(4-Trifluorophenyl)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2509906

  SDS
alfa aesar™
B2509906
5g Each for $203.42

AAB2509914

  SDS
alfa aesar™
B2509914
25g Each for $800.09
Description & Specifications

Specifications

Formula Weight 266.22
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
CAS 84392-17-6
Melting Point 167° to 170°C
Quantity 5g
Health Hazard 1 Warning
Chemical Name or Material 4'-(Trifluoromethyl)biphenyl-2-carboxylic acid
MDL Number MFCD00075353
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 7577560
Synonym 4-(4-Trifluorophenyl)benzoic acid
Molecular Formula C14H9F3O2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.