missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Trifluoromethyl)benzyl Chloride 97.0+%, TCI America™
Supplier: TCI America T24885G
Specifications
4-(Trifluoromethyl)benzyl Chloride | |
19°C | |
68°C | |
MFCD00040772 | |
3265 | |
MCHDHQVROPEJJT-UHFFFAOYSA-N | |
1-(chloromethyl)-4-(trifluoromethyl)benzene | |
123096 | |
≥97.0% (GC) |
939-99-1 | |
Yellow | |
C8H6ClF3 | |
5 g | |
4-trifluoromethyl benzyl chloride, 4-trifluoromethylbenzyl chloride, 1-chloromethyl-4-trifluoromethyl benzene, p-trifluoromethylbenzyl chloride, ccris 5110, 4-chloromethyl benzotrifluoride, p-trifluoromethyl benzyl chloride, benzene, 1-chloromethyl-4-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-p-xylene | |
C1=CC(=CC=C1CCl)C(F)(F)F | |
194.581 | |
194.58 | |
Liquid |
Chemical Identifiers
939-99-1 | |
194.581 | |
MCHDHQVROPEJJT-UHFFFAOYSA-N | |
123096 | |
C1=CC(=CC=C1CCl)C(F)(F)F |
C8H6ClF3 | |
MFCD00040772 | |
4-trifluoromethyl benzyl chloride, 4-trifluoromethylbenzyl chloride, 1-chloromethyl-4-trifluoromethyl benzene, p-trifluoromethylbenzyl chloride, ccris 5110, 4-chloromethyl benzotrifluoride, p-trifluoromethyl benzyl chloride, benzene, 1-chloromethyl-4-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-p-xylene | |
1-(chloromethyl)-4-(trifluoromethyl)benzene |
Safety and Handling
TSCA : No