missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)benzyl Alcohol, TCI America™

Manufacturer:  TCI America T131225G

 View more versions of this product

Catalog No. T131225G



Specifications

Specifications

4-(Trifluoromethyl)benzyl Alcohol
White-Yellow
349-95-1
MFCD00004661
MOOUWXDQAUXZRG-UHFFFAOYSA-N
[4-(trifluoromethyl)phenyl]methanol
67684
Crystal-Lump at 20°C
80°C
25g
C8H7F3O
4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol
C1=CC(=CC=C1CO)C(F)(F)F
176.138
176.14
≥96.0% (GC)

Chemical Identifiers

349-95-1
176.138
MOOUWXDQAUXZRG-UHFFFAOYSA-N
67684
C1=CC(=CC=C1CO)C(F)(F)F
C8H7F3O
MFCD00004661
4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol
[4-(trifluoromethyl)phenyl]methanol
Safety and Handling

Safety and Handling

TSCA : No

Documents