Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)benzyl alcohol, 99%, ACROS Organics™

$74.14 - $74.14

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)benzyl alcohol
Name Note 99%
CAS 349-95-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H7F3O
Linear Formula CF3C6H4CH2OH
Formula Weight 176.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC213570050

  SDS
fisher scientific acros
213570050
Glass bottle 5g Each for $74.14
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid After Melting
Formula Weight 176.14
Quantity 5g
Color Clear colorless to light brown
EINECS Number 206-494-6
Density 1.28g/mL
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Molecular Formula C8H7F3O
Linear Formula CF3C6H4CH2OH
MDL Number MFCD00004661
Name Note 99%
Chemical Name or Material 4-(Trifluoromethyl)benzyl alcohol
Assay Percent Range 99%
CAS 349-95-1
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.