Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethyl)benzophenone, 98%

$93.43 - $438.69

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)benzophenone
CAS 728-86-9
Molecular Formula C14H9F3O
Formula Weight 250.22
MDL Number MFCD00000400
Beilstein 1879572
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2077514

  SDS
alfa aesar™
B2077514
25g Each for $438.69

AAB2077506

  SDS
alfa aesar™
B2077506
5g Each for $93.43
Description & Specifications

Specifications

Formula Weight 250.22
Chemical Name or Material 4-(Trifluoromethyl)benzophenone
Odor Odorless
Assay 98%
Molecular Formula C14H9F3O
MDL Number MFCD00000400
CAS 728-86-9
Beilstein 1879572
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 116° to 119°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.