Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)benzophenone, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC214420010

 View more versions of this product

Catalog No. AC214420010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Solid
Formula Weight 250.22
Assay ≥96.0%
Quantity 1g
Color Off-white to beige
EINECS Number 211-974-3
Melting Point 113° to 116°C
Beilstein 07, IV, 1404
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-(Trifluoromethyl)benzophenone
Name Note 97%
Assay Percent Range 97%
Molecular Formula C14H9F3O
Linear Formula CF3C6H4COC6H5
MDL Number MFCD00000400
CAS 728-86-9
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-(Trifluoromethyl)benzophenone
Name Note 97%
CAS 728-86-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14H9F3O
Linear Formula CF3C6H4COC6H5
Formula Weight 250.22
MDL Number MFCD00000400
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents