Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)benzophenone, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)benzophenone
Name Note 97%
CAS 728-86-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14H9F3O
Linear Formula CF3C6H4COC6H5
Formula Weight 250.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC214420050

  SDS
thermo fisher scientific
214420050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Solid
Formula Weight 250.22
Quantity 5g
Color Off-white to beige
Beilstein 07, IV, 1404
EINECS Number 211-974-3
Melting Point 113° to 116°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Name Note 97%
Chemical Name or Material 4-(Trifluoromethyl)benzophenone
Linear Formula CF3C6H4COC6H5
Molecular Formula C14H9F3O
MDL Number MFCD00000400
CAS 728-86-9
Assay Percent Range 97%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.