missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)benzenesulfonamide, TCI America™

Manufacturer:  TCI America T284725G

 View more versions of this product

Catalog No. T284725G



Specifications

Specifications

4-(Trifluoromethyl)benzenesulfonamide
181°C
830-43-3
MFCD00159251
TVHXQQJDMHKGGK-UHFFFAOYSA-N
4-(trifluoromethyl)benzenesulfonamide
70018
Crystal-Powder at 20°C
White
25g
C7H6F3NO2S
4-trifluoromethyl benzenesulfonamide, 4-trifluoromethyl benzene-1-sulfonamide, benzenesulfonamide, p-trifluoromethyl, 4-trifluoromethylbenzenesulphonamide, p-trifluoromethyl benzenesulfonamide, 4-trifluoromethyl benzenesulphonamide, benzenesulfonamide, 4-trifluoromethyl, p-trifluoromethylbenzenesulfonamide, alpha,alpha,alpha-trifluoro-p-toluenesulphonamide, pubchem11767
C1=CC(=CC=C1C(F)(F)F)S(=O)(=O)N
225.185
225.19
≥98.0% (HPLC,N)

Chemical Identifiers

830-43-3
225.185
TVHXQQJDMHKGGK-UHFFFAOYSA-N
70018
C1=CC(=CC=C1C(F)(F)F)S(=O)(=O)N
C7H6F3NO2S
MFCD00159251
4-trifluoromethyl benzenesulfonamide, 4-trifluoromethyl benzene-1-sulfonamide, benzenesulfonamide, p-trifluoromethyl, 4-trifluoromethylbenzenesulphonamide, p-trifluoromethyl benzenesulfonamide, 4-trifluoromethyl benzenesulphonamide, benzenesulfonamide, 4-trifluoromethyl, p-trifluoromethylbenzenesulfonamide, alpha,alpha,alpha-trifluoro-p-toluenesulphonamide, pubchem11767
4-(trifluoromethyl)benzenesulfonamide
Safety and Handling

Safety and Handling

RTECSNumber : DB3200000

TSCA : No

Documents