missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2237406

 View more versions of this product

Catalog No. AAB2237406



Description

Description

4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.

Solubility
Soluble in DMSO, not in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

4-(Trifluoromethyl)benzeneboronic acid
238°C to 242°C
MFCD00151855
3544189
Soluble in DMSO,not in water
OB(O)C1=CC=C(C=C1)C(F)(F)F
189.93
189.93
128796-39-4
C7H6BF3O2
5 g
4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid
ALMFIOZYDASRRC-UHFFFAOYSA-N
[4-(trifluoromethyl)phenyl]boronic acid
2734389
98%

Chemical Identifiers

128796-39-4
189.93
ALMFIOZYDASRRC-UHFFFAOYSA-N
2734389
OB(O)C1=CC=C(C=C1)C(F)(F)F
C7H6BF3O2
MFCD00151855
4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid
[4-(trifluoromethyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only