Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)benzene-1,2-diamine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB07340DA

 View more versions of this product

Catalog No. BTB07340DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 176.14
Quantity 1g
Assay 95%
CAS Max % 100.0
Chemical Name or Material 4-(Trifluoromethyl)benzene-1, 2-diamine
Assay Percent Range 95%
Molecular Formula C7H7F3N2
CAS 368-71-8
CAS Min % 94.5
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
MDL Number MFCD00042456
Health Hazard 1 GHS Signal Word: Warning
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-(Trifluoromethyl)benzene-1, 2-diamine
CAS 368-71-8
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H7F3N2
Formula Weight 176.14
MDL Number MFCD00042456
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

SDS & Safety Documents