Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethyl)benzamide, 98+%

$93.43 - $93.43

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)benzamide
CAS 1891-90-3
Molecular Formula C8H6F3NO
Formula Weight 189.14
MDL Number MFCD00007998
Beilstein 2098929
Synonym alpha,alpha,alpha-Trifluoro-p-toluamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1274606

 
alfa aesar™
A1274606
5g Each for $93.43
Description & Specifications

Specifications

Formula Weight 189.14
Chemical Name or Material 4-(Trifluoromethyl)benzamide
Odor Odorless
Assay ≥98%
Molecular Formula C8H6F3NO
MDL Number MFCD00007998
CAS 1891-90-3
Beilstein 2098929
Synonym alpha,alpha,alpha-Trifluoro-p-toluamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 182° to 186°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.