missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethyl)benzaldehyde, 97%

Manufacturer:  Alfa Aesar™ A1527614

 View more versions of this product

Catalog No. AAA1527614



Description

Description

Applications
4-(Trifluoromethyl)benzaldehyde is useful reagent in kinetic studies of the asymmetric synthesis of alcohols and of the Wittig reaction. It is also used as Pharmaceutical intermediates.

Solubility
Soluble in water. 1.5 g/L at 20°C

Notes
Air Sensitive. Store away from air and oxidizing agents. Store under an inert atmosphere. Incompatible with oxidizing agents, reducing agents.
Specifications

Specifications

4-(Trifluoromethyl)benzaldehyde
80°C to 81°C (25mmHg)
25g
C8H5F3O
1101680
BEOBZEOPTQQELP-UHFFFAOYSA-N
4-(trifluoromethyl)benzaldehyde
67996
97%
65°C (149°F)
1°C to 2°C
455-19-6
MFCD00006952
4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl benzaldehyde, 4-trifluoromethyl-benzaldehyde, 4-trifluoromethyl benzaldehyde
C1=CC(=CC=C1C=O)C(F)(F)F
174.122
174.12

Chemical Identifiers

455-19-6
174.122
BEOBZEOPTQQELP-UHFFFAOYSA-N
67996
C1=CC(=CC=C1C=O)C(F)(F)F
C8H5F3O
MFCD00006952
4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl benzaldehyde, 4-trifluoromethyl-benzaldehyde, 4-trifluoromethyl benzaldehyde
4-(trifluoromethyl)benzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

TSCA : TSCA

Documents