Learn More
4-(Trifluoromethyl)aniline hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1832914
Description
4-(Trifluoromethyl)aniline hydrochloride is used in the synthesis of 2-(4-((6-Chloro-2-quinoxalinyl)oxy)phenoxy)-N-(4-(trifluoromethyl)phenyl)propanamide. It is also used to produce N-(p-trifluoromethylphenyl)phosphoramidic dichloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Trifluoromethyl)aniline hydrochloride is used in the synthesis of 2-(4-((6-Chloro-2-quinoxalinyl)oxy)phenoxy)-N-(4-(trifluoromethyl)phenyl)propanamide. It is also used to produce N-(p-trifluoromethylphenyl)phosphoramidic dichloride.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
Specifications
4-(Trifluoromethyl)aniline hydrochloride | |
∼240°C (decomposition) | |
MFCD00007897 | |
UN2811 | |
4-aminobenzotrifluoride hydrochloride, 4-trifluoromethyl aniline hydrochloride, 4-trifluoromethyl anilinium chloride, p-trifluoromethylaniline hydrochloride, 4-trifluoromethyl phenylamine, chloride, pubchem2816, 4-aminobenzotrifluoride, hydrochloride, 4-trifluoromethylaniline hydrochloride, 4-trifluoromethyl aniline;hydrochloride, a,a,a-trifluoro-p-toluidine hydrochloride | |
C1=CC(=CC=C1C(F)(F)F)N.Cl | |
197.585 | |
197.59 |
90774-69-9 | |
C7H7ClF3N | |
25 g | |
3629991 | |
CZGDYZDUMJDQSD-UHFFFAOYSA-N | |
4-(trifluoromethyl)aniline;hydrochloride | |
2723961 | |
97% |
Chemical Identifiers
90774-69-9 | |
197.585 | |
CZGDYZDUMJDQSD-UHFFFAOYSA-N | |
2723961 | |
C1=CC(=CC=C1C(F)(F)F)N.Cl |
C7H7ClF3N | |
MFCD00007897 | |
4-aminobenzotrifluoride hydrochloride, 4-trifluoromethyl aniline hydrochloride, 4-trifluoromethyl anilinium chloride, p-trifluoromethylaniline hydrochloride, 4-trifluoromethyl phenylamine, chloride, pubchem2816, 4-aminobenzotrifluoride, hydrochloride, 4-trifluoromethylaniline hydrochloride, 4-trifluoromethyl aniline;hydrochloride, a,a,a-trifluoro-p-toluidine hydrochloride | |
4-(trifluoromethyl)aniline;hydrochloride |
Safety and Handling
GHS H Statement
H301-H373-H315-H319-H335
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 292-679-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only