missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Trifluoromethyl)acetophenone, 98+%

Manufacturer:  Alfa Aesar™ A1552922

 View more versions of this product

Catalog No. AAA1552922



Specifications

Specifications

4'-(Trifluoromethyl)acetophenone
Characteristic
29°C to 35°C
709-63-7
MFCD00000401
4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl
CC(=O)C1=CC=C(C=C1)C(F)(F)F
188.149
188.15
84°C (183°F)
83°C to 85°C (9mmHg)
100g
C9H7F3O
1870425
HHAISVSEJFEWBZ-UHFFFAOYSA-N
1-[4-(trifluoromethyl)phenyl]ethanone
69731
≥98%

Chemical Identifiers

709-63-7
188.149
HHAISVSEJFEWBZ-UHFFFAOYSA-N
69731
CC(=O)C1=CC=C(C=C1)C(F)(F)F
C9H7F3O
MFCD00000401
4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl
1-[4-(trifluoromethyl)phenyl]ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents