missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Trifluoromethyl-2(1H)-quinolinone, 97%

Manufacturer:  Alfa Aesar™ H61732MD

 View more versions of this product

Catalog No. AAH61732MD



Specifications

Specifications

4-Trifluoromethyl-2(1H)-quinolinone
250mg
C10H6F3NO
2-hydroxy-4-trifluoromethyl quinoline, 4-trifluoromethyl quinolin-2-ol, 4-trifluoromethyl-2 1h-quinolinone, 4-trifluoromethyl-1h-quinolin-2-one, 2 1h-quinolinone, 4-trifluoromethyl, 4-trifluoromethyl quinolin-2 1h-one, pubchem5867, ksc495g1t, 4-trifluoromethylquinolin-2-one, 4-trifluoromethyl-quinolin-2-ol
C1=CC=C2C(=C1)C(=CC(=O)N2)C(F)(F)F
213.159
213.16
97%
White
25199-84-2
MFCD03407380
UUROBWTVZZNDFD-UHFFFAOYSA-N
4-(trifluoromethyl)-1H-quinolin-2-one
2759347
Solid

Chemical Identifiers

25199-84-2
213.159
UUROBWTVZZNDFD-UHFFFAOYSA-N
2759347
C1=CC=C2C(=C1)C(=CC(=O)N2)C(F)(F)F
C10H6F3NO
MFCD03407380
2-hydroxy-4-trifluoromethyl quinoline, 4-trifluoromethyl quinolin-2-ol, 4-trifluoromethyl-2 1h-quinolinone, 4-trifluoromethyl-1h-quinolin-2-one, 2 1h-quinolinone, 4-trifluoromethyl, 4-trifluoromethyl quinolin-2 1h-one, pubchem5867, ksc495g1t, 4-trifluoromethylquinolin-2-one, 4-trifluoromethyl-quinolin-2-ol
4-(trifluoromethyl)-1H-quinolin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

Documents