missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Trifluoromethoxyphenylboronic acid, 98%, ACROS Organics™

Chemical Identifiers

CAS 139301-27-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H6BF3O3
Formula Weight 205.93
MDL Number MFCD01074648
Chemical Name or Material 4-Trifluoromethoxyphenylboronic acid
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC359730050 SDS Thermo Fisher Scientific
AC359730050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC359730010 SDS Fisher Scientific Acros
35973-0010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

139301-27-2
100.0
C7H6BF3O3
MFCD01074648
≥97.5%
97.5
98%
205.93
4-Trifluoromethoxyphenylboronic acid

Specifications

Crystalline Powder
5g
100.0
Glass bottle
97.5
98%
4-Trifluoromethoxyphenylboronic acid
205.93
White to beige
C7H6BF3O3
MFCD01074648
139301-27-2
Authentic
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.