Please login to your online account to display your discounted pricing

4-(Trifluoromethoxy)phenylacetonitrile, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)phenylacetonitrile
CAS 49561-96-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H6F3NO
Linear Formula F3COC6H4CH2CN
Formula Weight 201.15
MDL Number MFCD00061240
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC379390010

  SDS
thermo fisher scientific
AC379390010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 201.15
Quantity 1g
Color Clear yellow
Density 1.2800g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Refractive Index 1.4480 to 1.4500 (20°C, 589nm)
CAS 49561-96-8
Health Hazard 1 Warning
Chemical Name or Material 4-(Trifluoromethoxy)phenylacetonitrile
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C9H6F3NO
Linear Formula F3COC6H4CH2CN
MDL Number MFCD00061240
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.