Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)phenylacetonitrile, 98%

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)phenylacetonitrile
CAS 49561-96-8
Molecular Formula C9H6F3NO
Formula Weight 201.15
MDL Number MFCD00061240
UN Number UN3439
Beilstein 2643789
Synonym 4-(Trifluoromethoxy)benzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2026703

  SDS
alfa aesar™
B2026703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2026706

  SDS
alfa aesar™
B2026706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.15
Chemical Name or Material 4-(Trifluoromethoxy)phenylacetonitrile
Assay 98%
Molecular Formula C9H6F3NO
MDL Number MFCD00061240
CAS 49561-96-8
UN Number UN3439
Beilstein 2643789
Synonym 4-(Trifluoromethoxy)benzyl cyanide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Flash Point >110°C (230°F)
Melting Point 28° to 30°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.