missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethoxy)phenylacetic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1951203

 View more versions of this product

Catalog No. AAA1951203



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Trifluoromethoxy)phenylacetic acid
86°C to 88°C
MFCD00061239
2695336
LQDBIEHCXRQVQI-LYAINCPBSA-L
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-[[4-[4-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]phenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
44135513
98%
4315-07-5
C34H22N8Na2O10S2
1 g
diamine green b, c.i. direct green 6, disodium salt, pontamine green, dianil green, 2,7-naphthalenedisulfonic acid, 4-amino-5-hydroxy-6-2-4'-2-4-hydroxyphenyl diazenyl 1,1'-biphenyl-4-yl diazenyl-3-2-4-nitrophenyl diazenyl-, sodium salt 1:2
C1=CC(=O)C=CC1=NNC2=CC=C(C=C2)C3=CC=C(C=C3)NN=C4C(=CC5=CC(=C(C(=C5C4=O)N)N=NC6=CC=C(C=C6)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
812.70
220.15

Chemical Identifiers

4315-07-5
MFCD00061239
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only