Please login to your online account to display your discounted pricing

4-(Trifluoromethoxy)phenyl isothiocyanate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)phenyl isothiocyanate
CAS 64285-95-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H4F3NOS
Linear Formula CF3OC6H4NCS
Formula Weight 219.19
MDL Number MFCD00042412
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00411EA

 
fisher scientific company
TL00411EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 219.19
Quantity 10g
Assay 97%
Density 1.3500g/mL
CAS Min % 97.0
Chemical Name or Material 4-(Trifluoromethoxy)phenyl isothiocyanate
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C8H4F3NOS
Linear Formula CF3OC6H4NCS
MDL Number MFCD00042412
CAS 64285-95-6
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.