Learn More
4-(Trifluoromethoxy)phenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6401303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(Trifluoromethoxy)phenyl isothiocyanate | |
Colorless | |
215°C to 218°C (literature) | |
C8H4F3NOS | |
MFCD00042412 | |
UN1760 | |
Moisture sensitive | |
JKSZUQPHKOPVHF-UHFFFAOYSA-N | |
1-isothiocyanato-4-(trifluoromethoxy)benzene | |
2777338 | |
97% |
64285-95-6 | |
1.35 g/mL | |
107°C (225°F) | |
1.541 | |
1 g | |
2723939 | |
4-trifluoromethoxy phenyl isothiocyanate, 1-isothiocyanato-4-trifluoromethoxy benzene, benzene,1-isothiocyanato-4-trifluoromethoxy, benzene, 1-isothiocyanato-4-trifluoromethoxy, 4-trifluoromethoxy phenylisothiocyanate, 1-isothiocyanato-4-trifluoromethoxy-benzene, acmc-1b5bx, 4-trifluoromethoxyphenyl isothiocyanate, 4-trifluoromethoxyphenyl-isothiocyanate, 4-trifluoromethoxy benzenisothiocyanate | |
C1=CC(=CC=C1N=C=S)OC(F)(F)F | |
219.181 | |
219.18 | |
Liquid |
Chemical Identifiers
64285-95-6 | |
219.181 | |
JKSZUQPHKOPVHF-UHFFFAOYSA-N | |
1-isothiocyanato-4-(trifluoromethoxy)benzene |
C8H4F3NOS | |
MFCD00042412 | |
2777338 | |
C1=CC(=CC=C1N=C=S)OC(F)(F)F |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only