Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)phenoxyacetic acid, 95%

$123.55 - $534.91

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)phenoxyacetic acid
CAS 72220-50-9
Molecular Formula C9H7F3O4
Formula Weight 236.15
MDL Number MFCD00052352
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5021803

  SDS
alfa aesar™
H5021803
1g Each for $123.55

AAH5021806

  SDS
alfa aesar™
H5021806
5g Each for $534.91
Description & Specifications

Specifications

Formula Weight 236.15
MDL Number MFCD00052352
Assay 95%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C9H7F3O4
Quantity 1g
Chemical Name or Material 4-(Trifluoromethoxy)phenoxyacetic acid
CAS 72220-50-9
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.