missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethoxy)phenol, TCI America™

Manufacturer:  TCI America T134125G

 View more versions of this product

Catalog No. T134125G



Specifications

Specifications

4-(Trifluoromethoxy)phenol
Yellow
828-27-3
MFCD00040988
WDRJNKMAZMEYOF-UHFFFAOYSA-N
4-(trifluoromethoxy)phenol
70015
Clear Liquid at 20°C
95°C
25g
C7H5F3O2
4-trifluoromethoxy phenol, p-trifluoromethoxy phenol, p-trifluoromethoxy phenol, phenol, 4-trifluoromethoxy, phenol, 4-trifluoromethoxy, 4-hydroxytrifluoromethoxybenzene, 4-trifluoromethoxy phenol, 4-trifluoromethoxy-phenol, 4-hydroxyphenyl trifluoromethyl ether, p-trifluoromethoxyphenol
C1=CC(=CC=C1O)OC(F)(F)F
178.11
178.11
≥95.0% (GC)

Chemical Identifiers

828-27-3
178.11
WDRJNKMAZMEYOF-UHFFFAOYSA-N
70015
C1=CC(=CC=C1O)OC(F)(F)F
C7H5F3O2
MFCD00040988
4-trifluoromethoxy phenol, p-trifluoromethoxy phenol, p-trifluoromethoxy phenol, phenol, 4-trifluoromethoxy, phenol, 4-trifluoromethoxy, 4-hydroxytrifluoromethoxybenzene, 4-trifluoromethoxy phenol, 4-trifluoromethoxy-phenol, 4-hydroxyphenyl trifluoromethyl ether, p-trifluoromethoxyphenol
4-(trifluoromethoxy)phenol
Safety and Handling

Safety and Handling

TSCA : No

Documents