Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)phenol, 98%

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)phenol
CAS 828-27-3
Molecular Formula C7H5F3O2
Formula Weight 178.11
MDL Number MFCD00040988
UN Number UN2810
Beilstein 1945934
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1269503

 
alfa aesar™
A1269503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1269506

 
alfa aesar™
A1269506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1269514

 
alfa aesar™
A1269514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.11
Chemical Name or Material 4-(Trifluoromethoxy)phenol
Assay 98%
Molecular Formula C7H5F3O2
MDL Number MFCD00040988
CAS 828-27-3
UN Number UN2810
Beilstein 1945934
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H318-H227-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Flash Point 86°C (186°F)
Boiling Point 91° to 92°C (25mmHg)
Melting Point 17° to 18°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H227-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.