Learn More
4-(Trifluoromethoxy)iodobenzene, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 443410010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-(Trifluoromethoxy)iodobenzene | |
96.0 | |
1.8400g/mL | |
78°C | |
96% min. (GC) | |
C7H4F3IO | |
IC6H4OCF3 | |
1g | |
1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene | |
FC(F)(F)OC1=CC=C(I)C=C1 | |
288.01 | |
288 |
103962-05-6 | |
100.0 | |
72.0°C to 74.0°C (15.0mmHg) | |
Authentic | |
Glass Bottle | |
1.504 | |
MFCD00042411 | |
1.84 | |
RTUDBROGOZBBIC-UHFFFAOYSA-N | |
1-iodo-4-(trifluoromethoxy)benzene | |
2777294 | |
97% |
Chemical Identifiers
103962-05-6 | |
288.01 | |
RTUDBROGOZBBIC-UHFFFAOYSA-N | |
2777294 | |
FC(F)(F)OC1=CC=C(I)C=C1 |
C7H4F3IO | |
MFCD00042411 | |
1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene | |
1-iodo-4-(trifluoromethoxy)benzene |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
Warning
RUO â Research Use Only