Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)-DL-phenylglycine, 97%

$80.61 - $266.70

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)-DL-phenylglycine
CAS 261952-24-3
Molecular Formula C9H8NO3F3
Formula Weight 235.16
MDL Number MFCD01631461
Synonym 2-Amino-2-(4-trifluoromethoxyphenyl)acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3124503

  SDS
alfa aesar™
H3124503
1g Each for $266.70

AAH31245MD

  SDS
alfa aesar™
H31245MD
250mg Each for $80.61
Description & Specifications

Specifications

Formula Weight 235.16
CAS 261952-24-3
Synonym 2-Amino-2-(4-trifluoromethoxyphenyl)acetic acid
Quantity 1g
Chemical Name or Material 4-(Trifluoromethoxy)-DL-phenylglycine
Assay 97%
Molecular Formula C9H8NO3F3
MDL Number MFCD01631461
Melting Point ≈292°C (subl.)