Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)benzylamine, 97%

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)benzylamine
CAS 93919-56-3
Molecular Formula C8H8F3NO
Formula Weight 191.15
MDL Number MFCD00061237
UN Number UN2735
Beilstein 8200624
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1802903

  SDS
alfa aesar™
A1802903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1802906

  SDS
alfa aesar™
A1802906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1802914

  SDS
alfa aesar™
A1802914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.15
Chemical Name or Material 4-(Trifluoromethoxy)benzylamine
Assay 97%
Molecular Formula C8H8F3NO
MDL Number MFCD00061237
CAS 93919-56-3
UN Number UN2735
Beilstein 8200624
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Flash Point 77°C (170°F)
Boiling Point 57° to 60°C (10mmHg)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.