missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethoxy)benzyl bromide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 293510025

 View more versions of this product

Catalog No. AC293510025



Description

Description

Specifications

Specifications

4-(Trifluoromethoxy)benzyl bromide
94°C
Glass bottle
1.58
Yellow
2.5g
97.5
97.5% min. (GC)
CF3OC6H4CH2Br
1-bromomethyl-4-trifluoromethoxy benzene, 1-bromomethyl-4-trifluoromethoxy-benzene, 4-bromomethyl phenoxy trifluoromethane, 4-trifluoromethoxy benzyl bromide, 4-trifluoromethoxy benzylbromide, 4-trifluoromethoxy-benzyl bromide, 4-trifluoromethoxybenzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, p-trifluoromethoxy benzyl bromide
C1=CC(=CC=C1CBr)OC(F)(F)F
255.03
255.03
98%
1.5800g/mL
Authentic
1.4800 to 1.482
82°C to 84°C (10.0mmHg)
23°C to 24°C
50824-05-0
100.0
C8H6BrF3O
MFCD00061238
JDNPUJCKXLOHOW-UHFFFAOYSA-N
1-(bromomethyl)-4-(trifluoromethoxy)benzene
142785
Liquid or Low Melting Solid

Chemical Identifiers

50824-05-0
255.03
JDNPUJCKXLOHOW-UHFFFAOYSA-N
142785
C1=CC(=CC=C1CBr)OC(F)(F)F
C8H6BrF3O
MFCD00061238
1-bromomethyl-4-trifluoromethoxy benzene, 1-bromomethyl-4-trifluoromethoxy-benzene, 4-bromomethyl phenoxy trifluoromethane, 4-trifluoromethoxy benzyl bromide, 4-trifluoromethoxy benzylbromide, 4-trifluoromethoxy-benzyl bromide, 4-trifluoromethoxybenzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, p-trifluoromethoxy benzyl bromide
1-(bromomethyl)-4-(trifluoromethoxy)benzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents