Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)benzyl alcohol, 97%

$52.49 - $167.78

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)benzyl alcohol
CAS 1736-74-9
Molecular Formula C8H7F3O2
Formula Weight 192.14
MDL Number MFCD00036029
Beilstein 1950379
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1925003

  SDS
alfa aesar™
A1925003
1g Each for $52.49

AAA1925006

  SDS
alfa aesar™
A1925006
5g Each for $167.78
Description & Specifications

Specifications

Formula Weight 192.14
Chemical Name or Material 4-(Trifluoromethoxy)benzyl alcohol
Assay 97%
Molecular Formula C8H7F3O2
MDL Number MFCD00036029
CAS 1736-74-9
Beilstein 1950379
Flash Point 98°C (208°F)
Boiling Point 100° to 102°C (12mmHg)
Quantity 1g