Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)benzotrifluoride, 97%

$39.18 - $153.69

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)benzotrifluoride
CAS 80258-33-9
Molecular Formula C8H4F6O
Formula Weight 230.11
MDL Number MFCD00190125
Beilstein 6206351
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2453403

  SDS
alfa aesar™
B2453403
1g Each for $39.18

AAB2453406

  SDS
alfa aesar™
B2453406
5g Each for $153.69
Description & Specifications

Specifications

Formula Weight 230.11
Assay 97%
MDL Number MFCD00190125
Health Hazard 1 Warning
CAS 80258-33-9
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H4F6O
Chemical Name or Material 4-(Trifluoromethoxy)benzotrifluoride
Beilstein 6206351
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.