Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)benzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)benzonitrile
CAS 332-25-2
Molecular Formula C8H4F3NO
Formula Weight 187.12
MDL Number MFCD00039474
UN Number UN3276
Beilstein 2832939
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1312403

  SDS
alfa aesar™
A1312403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1312406

  SDS
alfa aesar™
A1312406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.12
Chemical Name or Material 4-(Trifluoromethoxy)benzonitrile
Assay 98%
Molecular Formula C8H4F3NO
MDL Number MFCD00039474
CAS 332-25-2
UN Number UN3276
Beilstein 2832939
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Flash Point 82°C (179°F)
Boiling Point 195° to 196°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.