Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)benzoic acid, 97%

$56.50 - $854.34

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)benzoic acid
CAS 330-12-1
Molecular Formula C8H5F3O3
Formula Weight 206.12
MDL Number MFCD00002541
Beilstein 977356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1362103

  SDS
alfa aesar™
A1362103
1g Each for $56.50

AAA1362114

  SDS
alfa aesar™
A1362114
25g Each for $854.34

AAA1362106

  SDS
alfa aesar™
A1362106
5g Each for $201.91
Description & Specifications

Specifications

Formula Weight 206.12
Molecular Formula C8H5F3O3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
MDL Number MFCD00002541
Quantity 1g
CAS 330-12-1
Health Hazard 1 Warning
Melting Point 151° to 154°C
Chemical Name or Material 4-(Trifluoromethoxy)benzoic acid
Beilstein 977356
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.