missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethoxy)benzenesulfonamide, TCI America™

Manufacturer:  TCI America T242725G

 View more versions of this product

Catalog No. T242725G



Specifications

Specifications

4-(Trifluoromethoxy)benzenesulfonamide
149°C
1513-45-7
MFCD00798381
RGOJCHYYBKMRLL-UHFFFAOYSA-N
4-(trifluoromethoxy)benzenesulfonamide
2777211
Crystal-Powder at 20°C
White-Yellow
25g
C7H6F3NO3S
4-trifluoromethoxy benzenesulfonamide, p-trifluoromethoxy benzenesulfonamide, 4-trifluoromethoxy benzenesulphonamide, 4-trifluoromethoxy benzene-1-sulfonamide, benzenesulfonamide, 4-trifluoromethoxy, pubchem11787, rgojchyybkmrll-uhfffaoysa, 4-trifluoromethoxybenzensulfonamide, trifluoromethoxy benzenesulfonamide, 4-trifluoromethoxybenzenesulphonyl amine
C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)N
241.184
241.18
≥98.0% (HPLC,N)

Chemical Identifiers

1513-45-7
241.184
RGOJCHYYBKMRLL-UHFFFAOYSA-N
2777211
C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)N
C7H6F3NO3S
MFCD00798381
4-trifluoromethoxy benzenesulfonamide, p-trifluoromethoxy benzenesulfonamide, 4-trifluoromethoxy benzenesulphonamide, 4-trifluoromethoxy benzene-1-sulfonamide, benzenesulfonamide, 4-trifluoromethoxy, pubchem11787, rgojchyybkmrll-uhfffaoysa, 4-trifluoromethoxybenzensulfonamide, trifluoromethoxy benzenesulfonamide, 4-trifluoromethoxybenzenesulphonyl amine
4-(trifluoromethoxy)benzenesulfonamide
Safety and Handling

Safety and Handling

TSCA : No

Documents