Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)benzenesulfonamide, 97%

$94.31 - $345.05

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)benzenesulfonamide
CAS 1513-45-7
Molecular Formula C7H6F3NO3S
Formula Weight 241.19
MDL Number MFCD00798381
Beilstein 2653449
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1971603

  SDS
alfa aesar™
L1971603
1g Each for $94.31

AAL1971606

  SDS
alfa aesar™
L1971606
5g Each for $345.05
Description & Specifications

Specifications

Formula Weight 241.19
MDL Number MFCD00798381
CAS 1513-45-7
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H6F3NO3S
Melting Point 146° to 148°C
Quantity 1g
Chemical Name or Material 4-(Trifluoromethoxy)benzenesulfonamide
Beilstein 2653449
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.