Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)benzeneboronic acid, 98%

$65.09 - $861.86

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)benzeneboronic acid
CAS 139301-27-2
Molecular Formula C7H6BF3O3
Formula Weight 205.93
MDL Number MFCD01074648
Beilstein 8548201
Synonym 4-(Trifluoromethoxy)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2323303

  SDS
alfa aesar™
B2323303
1g Each for $65.09

AAB2323306

  SDS
alfa aesar™
B2323306
5g Each for $221.50

AAB2323314

  SDS
alfa aesar™
B2323314
25g Each for $861.86
Description & Specifications

Specifications

Formula Weight 205.93
Assay 98%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 8548201
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H6BF3O3
Synonym 4-(Trifluoromethoxy)phenylboronic acid
Quantity 1g
MDL Number MFCD01074648
CAS 139301-27-2
Chemical Name or Material 4-(Trifluoromethoxy)benzeneboronic acid
Melting Point ≈120° to 125°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.