Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethoxy)benzamide, 97%

$61.03 - $197.39

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethoxy)benzamide
CAS 456-71-3
Molecular Formula C8H6F3NO2
Formula Weight 205.14
MDL Number MFCD00041384
Beilstein 2837677
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1254803

  SDS
alfa aesar™
A1254803
1g Each for $61.03

AAA1254806

  SDS
alfa aesar™
A1254806
5g Each for $197.39
Description & Specifications

Specifications

Formula Weight 205.14
Assay 97%
Beilstein 2837677
Melting Point 155° to 156°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
MDL Number MFCD00041384
Chemical Name or Material 4-(Trifluoromethoxy)benzamide
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H6F3NO2
CAS 456-71-3
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.