missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethoxy)benzaldehyde, TCI America™

Manufacturer:  TCI America T225625G

 View more versions of this product

Catalog No. T225625G



Specifications

Specifications

4-(Trifluoromethoxy)benzaldehyde
Yellow
659-28-9
4-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, 4-trifluoromethoxy benzaldehyde, p-trifluoromethoxy benzaldehyde, benzaldehyde, 4-trifluoromethoxy, 4-trifluoromethoxy-benzaldehyde, 4-trifluoromethoxy-benzaldehyde, alpha,alpha,alpha-trifluoroanisaldehyde, pubchem1471, acmc-209nsx
C1=CC(=CC=C1C=O)OC(F)(F)F
190.121
190.12
≥97.0% (GC)
93°C
25g
C8H5F3O2
XQNVDQZWOBPLQZ-UHFFFAOYSA-N
4-(trifluoromethoxy)benzaldehyde
69573
Clear Liquid at 20°C

Chemical Identifiers

659-28-9
190.121
4-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, 4-trifluoromethoxy benzaldehyde, p-trifluoromethoxy benzaldehyde, benzaldehyde, 4-trifluoromethoxy, 4-trifluoromethoxy-benzaldehyde, 4-trifluoromethoxy-benzaldehyde, alpha,alpha,alpha-trifluoroanisaldehyde, pubchem1471, acmc-209nsx
4-(trifluoromethoxy)benzaldehyde
C8H5F3O2
XQNVDQZWOBPLQZ-UHFFFAOYSA-N
69573
C1=CC(=CC=C1C=O)OC(F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Documents