missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethoxy)benzaldehyde, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC293500010

 View more versions of this product

Catalog No. AC293500010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-(Trifluoromethoxy)benzaldehyde
70°C
Glass bottle
1.33
Colorless to Green-Yellow
659-28-9
100.0
C8H5F3O2
MFCD00041530
XQNVDQZWOBPLQZ-UHFFFAOYSA-N
4-(trifluoromethoxy)benzaldehyde
69573
Liquid
1.3300g/mL
Authentic
1.4570 to 1.459
80°C (16.0mmHg)
1g
98.5
98.5% min. (GC)
CF3OC6H4CHO
4-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy-benzaldehyde, acmc-209nsx, alpha,alpha,alpha-trifluoroanisaldehyde, benzaldehyde, 4-trifluoromethoxy, p-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, pubchem1471
C1=CC(=CC=C1C=O)OC(F)(F)F
190.12
190.12
99%

Chemical Identifiers

659-28-9
190.12
XQNVDQZWOBPLQZ-UHFFFAOYSA-N
69573
C1=CC(=CC=C1C=O)OC(F)(F)F
C8H5F3O2
MFCD00041530
4-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy-benzaldehyde, acmc-209nsx, alpha,alpha,alpha-trifluoroanisaldehyde, benzaldehyde, 4-trifluoromethoxy, p-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, pubchem1471
4-(trifluoromethoxy)benzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-531-4

SDS Documents