Please login to your online account to display your discounted pricing

4'-(Trifluoromethoxy)acetophenone, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC306230010

 View more versions of this product

Catalog No. AC306230010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5% (GC)
Formula Weight 204.15
Quantity 1g
Color Yellow to light brown
Density 1.2780g/mL
CAS Max % 100.0
Chemical Name or Material 4'-(Trifluoromethoxy)acetophenone
NMR Consistent with structure (1H-NMR)
Assay Percent Range 98%
Molecular Formula C9H7F3O2
MDL Number MFCD00042404
CAS 85013-98-5
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CF3OC6H4COCH3

Specifications

Chemical Name or Material 4'-(Trifluoromethoxy)acetophenone
CAS 85013-98-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C9H7F3O2
Linear Formula CF3OC6H4COCH3
Formula Weight 204.15
MDL Number MFCD00042404
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.