missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Trifluoromethoxy)acetophenone, 98%

$62.82 - $215.42

Chemical Identifiers

CAS 85013-98-5
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD00042404
Beilstein 5930486
Chemical Name or Material 4'-(Trifluoromethoxy)acetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1392803 SDS Alfa Aesar™
A1392803
1g Each for $62.82
AAA1392806 SDS Alfa Aesar™
A1392806
5g Each for $215.42
Specifications Description & Specifications

Chemical Identifiers

85013-98-5
204.15
5930486
C9H7F3O2
MFCD00042404
4'-(Trifluoromethoxy)acetophenone

Specifications

87°C (188°F)
1g
C9H7F3O2
MFCD00042404
4'-(Trifluoromethoxy)acetophenone
47° to 49°C (15mmHg)
85013-98-5
204.15
5930486
98%

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning