missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoroacetyl)benzoic acid, 97+%

Manufacturer:  Alfa Aesar™ L1024203

 View more versions of this product

Catalog No. AAL1024203



Specifications

Specifications

4-(Trifluoroacetyl)benzoic acid
1g
C9H5F3O3
2728778
WLTZCRCZDLLXQP-UHFFFAOYSA-N
4-(2,2,2-trifluoroacetyl)benzoic acid
143579
≥97%
177°C to 179°C
58808-59-6
MFCD00052340
4-trifluoroacetyl benzoic acid, 4-2,2,2-trifluoroacetyl benzoic acid, benzoic acid, 4-trifluoroacetyl, 4-2,2,2-trifluoro-acetyl-benzoic acid, benzoic acid, 4-trifluoroacetyl-9ci, 4-carboxy-a,a,a-trifluoroacetophenone, pubchem13997, acmc-1atst, 4-trifluoroacetylbenzoic acid, labotest-bb lt00454127
C1=CC(=CC=C1C(=O)C(F)(F)F)C(=O)O
218.131
218.13

Chemical Identifiers

58808-59-6
218.131
WLTZCRCZDLLXQP-UHFFFAOYSA-N
143579
C1=CC(=CC=C1C(=O)C(F)(F)F)C(=O)O
C9H5F3O3
MFCD00052340
4-trifluoroacetyl benzoic acid, 4-2,2,2-trifluoroacetyl benzoic acid, benzoic acid, 4-trifluoroacetyl, 4-2,2,2-trifluoro-acetyl-benzoic acid, benzoic acid, 4-trifluoroacetyl-9ci, 4-carboxy-a,a,a-trifluoroacetophenone, pubchem13997, acmc-1atst, 4-trifluoroacetylbenzoic acid, labotest-bb lt00454127
4-(2,2,2-trifluoroacetyl)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents