Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Tridecanone, 98%

$60.66 - $781.96

Chemical Identifiers

Chemical Name or Material 4-Tridecanone
CAS 26215-90-7
Molecular Formula CH3(CH2)8COCH2CH2CH3
Formula Weight 198.35
MDL Number MFCD00027351
Synonym n-Propyl n-nonyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2472103

 
alfa aesar™
B2472103
1g Each for $60.66

AAB2472106

 
alfa aesar™
B2472106
5g Each for $212.42

AAB2472114

 
alfa aesar™
B2472114
25g Each for $781.96
Description & Specifications

Specifications

Formula Weight 198.35
Chemical Name or Material 4-Tridecanone
Assay 98%
Molecular Formula CH3(CH2)8COCH2CH2CH3
MDL Number MFCD00027351
CAS 26215-90-7
Synonym n-Propyl n-nonyl ketone
Melting Point 18° to 19°C
Quantity 1g