missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-trans-Hydroxy-L-proline ethyl ester hydrochloride, 97%

$53.97 - $639.99

Chemical Identifiers

CAS 33996-30-4
Molecular Formula C7H14ClNO3
Molecular Weight (g/mol) 195.643
MDL Number MFCD00040589
InChI Key HHXSZDXMSRXWJV-IBTYICNHSA-N
Synonym h-hyp-oet hcl, h-hyp-oet.hcl, 2s,4r-ethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride, l-4-hydroxyproline ethyl ester hydrochloride, ethyl trans-4-hydroxy-l-prolinate hydrochloride, trans-4-hydroxy-l-proline ethyl ester hydrochloride, ethyl 2s,4r-4-hydroxypyrrolidine-2-carboxylate hydrochloride, ethyl trans-4-hydroxy-l-prolinate hcl, 2s,4r---ethyl 4-hydroxy-2-pyrrolidincarboxylate hydrochloride 99+%, 2s,4r-4-hydroxyproline ethyl ester hydrochloride
PubChem CID 12709304
IUPAC Name ethyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate;hydrochloride
SMILES CCOC(=O)C1CC(CN1)O.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6563306 View Documents Alfa Aesar™
H6563306
5g Each for $53.97
 
AAH6563314 View Documents Alfa Aesar™
H6563314
25g Each for $200.47
 
AAH6563322 View Documents Alfa Aesar™
H6563322
100g Each for $639.99
 
Specifications

Chemical Identifiers

33996-30-4
195.643
HHXSZDXMSRXWJV-IBTYICNHSA-N
12709304
CCOC(=O)C1CC(CN1)O.Cl
C7H14ClNO3
MFCD00040589
h-hyp-oet hcl, h-hyp-oet.hcl, 2s,4r-ethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride, l-4-hydroxyproline ethyl ester hydrochloride, ethyl trans-4-hydroxy-l-prolinate hydrochloride, trans-4-hydroxy-l-proline ethyl ester hydrochloride, ethyl 2s,4r-4-hydroxypyrrolidine-2-carboxylate hydrochloride, ethyl trans-4-hydroxy-l-prolinate hcl, 2s,4r---ethyl 4-hydroxy-2-pyrrolidincarboxylate hydrochloride 99+%, 2s,4r-4-hydroxyproline ethyl ester hydrochloride
ethyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate;hydrochloride

Specifications

33996-30-4
MFCD00040589
h-hyp-oet hcl, h-hyp-oet.hcl, 2s,4r-ethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride, l-4-hydroxyproline ethyl ester hydrochloride, ethyl trans-4-hydroxy-l-prolinate hydrochloride, trans-4-hydroxy-l-proline ethyl ester hydrochloride, ethyl 2s,4r-4-hydroxypyrrolidine-2-carboxylate hydrochloride, ethyl trans-4-hydroxy-l-prolinate hcl, 2s,4r---ethyl 4-hydroxy-2-pyrrolidincarboxylate hydrochloride 99+%, 2s,4r-4-hydroxyproline ethyl ester hydrochloride
CCOC(=O)C1CC(CN1)O.Cl
195.643
195.64
97%
5g
C7H14ClNO3
3656870
HHXSZDXMSRXWJV-IBTYICNHSA-N
ethyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate;hydrochloride
12709304
Powder
White
4-trans-Hydroxy-L-proline ethyl ester hydrochloride
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C