missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(trans-4-Propylcyclohexyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$54.06 - $54.06
Chemical Identifiers
| CAS | 146862-02-4 |
|---|---|
| Molecular Formula | C15H23BO2 |
| MDL Number | MFCD06658197 |
| InChI Key | QTBUVZSHCNAPRT-UHFFFAOYSA-N |
| PubChem CID | 3104710 |
| IUPAC Name | [4-(4-propylcyclohexyl)phenyl]boronic acid |
Chemical Identifiers
| 146862-02-4 | |
| MFCD06658197 | |
| 3104710 |
| C15H23BO2 | |
| QTBUVZSHCNAPRT-UHFFFAOYSA-N | |
| [4-(4-propylcyclohexyl)phenyl]boronic acid |
Specifications
| 146862-02-4 | |
| C15H23BO2 | |
| 1 g | |
| QTBUVZSHCNAPRT-UHFFFAOYSA-N | |
| [4-(4-propylcyclohexyl)phenyl]boronic acid | |
| 3104710 | |
| Crystalline Powder |
| White | |
| MFCD06658197 | |
| 4-trans-4-propylcyclohexyl phenylboronic acid, 4-trans-4-n-propylcyclohexyl phenyl boronic acid, 4-trans-4-propylcyclohexyl phenyl boronic acid, 4-4-propylcyclohexyl phenyl boronic acid, 4-4-propylcyclohexyl phenylboronic acid, boronic acid, 4-trans-4-propylcyclohexyl phenyl, 4-1s,4r-4-propylcyclohexyl phenylboronic acid, pubchem7891, 4-4-propylcyclohexyl phenylboronicacid | |
| B(C1=CC=C(C=C1)C2CCC(CC2)CCC)(O)O | |
| 246.157 | |
| 246.16 | |
| 4-(trans-4-Propylcyclohexyl)phenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No