missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(trans-3-Methoxy-3-oxo-1-propen-1-yl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H53292MD

 View more versions of this product

Catalog No. AAH53292MD



Description

Description

Applications
4-(trans-3-Methoxy-3-oxo-1-propen-1-yl)benzeneboronic acid used as reagents for suzuki-miyaura cross-coupling reacion.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate.
Specifications

Specifications

4-(trans-3-Methoxy-3-oxo-1-propen-1-yl)benzeneboronic acid
250mg
C10H11BO4
4-e-3-methoxy-3-oxo-1-propen-1-yl phenyl boronic acid, 4-3-methoxy-3-oxoprop-1-en-1-yl phenyl boronic acid, 4-e-3-methoxy-3-oxo-1-propen-1-yl phenylboronic acid, 4-trans-3-methoxy-3-oxo-1-propen-1-yl benzeneboronic acid, 4-3-methoxy-3-oxoprop-1-enyl phenylboronic acid, 4-1e-3-methoxy-3-oxoprop-1-en-1-yl phenylboronic acid, 4-3-methoxy-3-oxoprop-1-en-1-yl phenylboronic acid, 4-e-3-methoxy-3-oxoprop-1-enyl phenyl boronic acid, 4-1e-3-methoxy-3-oxoprop-1-en-1-yl phenyl boronic acid, 2-propenoic acid, 3-4-boronophenyl-, 1-methyl ester, 2e
B(C1=CC=C(C=C1)C=CC(=O)OC)(O)O
206.004
206
234°C to 240°C
380430-58-0
MFCD02179476
BIKAXBPCHWDATP-QPJJXVBHSA-N
[4-[(E)-3-methoxy-3-oxoprop-1-enyl]phenyl]boronic acid
5708389
97%

Chemical Identifiers

380430-58-0
206.004
BIKAXBPCHWDATP-QPJJXVBHSA-N
5708389
B(C1=CC=C(C=C1)C=CC(=O)OC)(O)O
C10H11BO4
MFCD02179476
4-e-3-methoxy-3-oxo-1-propen-1-yl phenyl boronic acid, 4-3-methoxy-3-oxoprop-1-en-1-yl phenyl boronic acid, 4-e-3-methoxy-3-oxo-1-propen-1-yl phenylboronic acid, 4-trans-3-methoxy-3-oxo-1-propen-1-yl benzeneboronic acid, 4-3-methoxy-3-oxoprop-1-enyl phenylboronic acid, 4-1e-3-methoxy-3-oxoprop-1-en-1-yl phenylboronic acid, 4-3-methoxy-3-oxoprop-1-en-1-yl phenylboronic acid, 4-e-3-methoxy-3-oxoprop-1-enyl phenyl boronic acid, 4-1e-3-methoxy-3-oxoprop-1-en-1-yl phenyl boronic acid, 2-propenoic acid, 3-4-boronophenyl-, 1-methyl ester, 2e
[4-[(E)-3-methoxy-3-oxoprop-1-enyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents