missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Tolylboronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 344430010

 View more versions of this product

Catalog No. AC344430010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Tolylboronic acid
96.0
256°C to 263°C
Authentic
Glass bottle
CH3C6H4B(OH)2
1 g
BIWQNIMLAISTBV-UHFFFAOYSA-N
(4-methylphenyl)boronic acid
79799
97%
5720-05-8
100.0
White
97%
C7H9BO2
MFCD00039138
4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid
CC1=CC=C(C=C1)B(O)O
135.96
135.96
Crystalline Powder

Chemical Identifiers

5720-05-8
135.96
BIWQNIMLAISTBV-UHFFFAOYSA-N
79799
CC1=CC=C(C=C1)B(O)O
C7H9BO2
MFCD00039138
4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid
(4-methylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physicia

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only