Please login to your online account to display your discounted pricing

4-(thien-2-ylmethyl)benzoyl chloride, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC70602DE

 View more versions of this product

Catalog No. CC70602DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 236.72
Quantity 5g
Assay 97%
Chemical Name or Material 4-(thien-2-ylmethyl)benzoyl chloride
Molecular Formula C12H9ClOS
CAS 115151219
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 4-(thien-2-ylmethyl)benzoyl chloride
Assay 97%
Molecular Formula C12H9ClOS
Formula Weight 236.72
Accessories

GHS Signal Word: Danger

SDS & Safety Documents