Please login to your online account to display your discounted pricing

4-(thien-2-ylmethyl)benzoic acid, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC70601CB

 View more versions of this product

Catalog No. CC70601CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 218.27
Quantity 250mg
Assay 97%
CAS 100272790
Molecular Formula C12H10O2S
Packaging Amber Glass Bottle
Chemical Name or Material 4-(thien-2-ylmethyl)benzoic acid

Specifications

Chemical Name or Material 4-(thien-2-ylmethyl)benzoic acid
Assay 97%
Molecular Formula C12H10O2S
Formula Weight 218.27