Please login to your online account to display your discounted pricing

4-(thien-2-ylmethyl)benzoic acid, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(thien-2-ylmethyl)benzoic acid
Assay 97%
Molecular Formula C12H10O2S
Formula Weight 218.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC70601DA

  SDS
fisher scientific company
CC70601DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC70601CB

  SDS
fisher scientific company
CC70601CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC70601DE

  SDS
fisher scientific company
CC70601DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.27
Quantity 1g
Assay 97%
CAS 100272790
Chemical Name or Material 4-(thien-2-ylmethyl)benzoic acid
Packaging Amber Glass Bottle
Molecular Formula C12H10O2S